Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS CIP 200™ Acid Detergent

Catalog No. 17463025 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
5 gal.
55 gal.
300 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-463-025 5 gal.
17-463-023 55 gal.
17-463-021 300 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-463-025 Supplier STERIS Supplier No. 1D2005
This item is not returnable. View return policy

The Acid-Based Process and Research Cleaner is a Phosphoric Acid Detergent for Routine Cleaning and Rouge Removal

CIP 200™ Acid-Based Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

 

HOW CIP 200 WORKS

CIP 200 Acid Detergent is a proprietary blend of phosphoric acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming detergent is ideal for use in CIP, COP and manual applications for a variety of applications including derouging and passivation.

 

WHY CIP 200?

Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
Global Availability - and manufactured with biodegradable surfactants to support harmonization of cleaning processes
Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Acid Detergent
pH 2
Appearance Colorless
Composition Proprietary Phosphoric Acid Blend with an Advanced Surfactant System
Container Type Jerrican
Solubility Complete
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.