Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ CIP Additive Detergent Booster and Defoamer

Catalog No. 17355274 Shop All STERIS Corporation Products
Change view
Click to view available options
Size:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-274 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-355-274 Supplier STERIS™ Supplier No. 1D0305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CIP Additive Detergent Booster and Defoamer is a neutral pH detergent additive for use in boosting the cleaning ability of either alkaline or acidic cleaners. CIP Additive Detergent Booster and Defoamer can be used to supplement surfactants and to enhance cleaning. pH - undiluted 8.57, typical.

CIP Additive Detergent Booster and Defoamer is specially formulated to enhance the cleaning performance of STERIS formulated detergents in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW CIP ADDITIVE WORKS

CIP Additive Detergent is a proprietary blend of surfactants and other performance-enhancing ingredients. This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.

WHY CIP ADDITIVE?

  • Productivity - Enhances the performance of other formulated detergents with additional sufactants.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Safety - Enhanced operator safety due to moderate pH.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Detergent Additive
Bristle Material No
Color-coded No
Form Clear Liquid
pH 8.57 (Undiluted)
Scent Unscented
Sterility Non-sterile
Size 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.