Learn More
Aviva Systems Biology Corp CIP29 Antibody - C-terminal region (ARP50729_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50729_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CIP29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CIP29. Alternate names: HCC-1, HCC1, HSPC316, MGC14726, THO1, CIP29ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60419