Learn More
ABclonal Technology Cip4 Rabbit mAb
SDP
Catalog No. 5000007934
Encompass_Preferred
Enables identical protein binding activity Predicted to be involved in actin cytoskeleton organization endocytosis and signal transduction Located in nucleoplasm Biomarker of Huntington s disease
Catalog No. 50-000-07934 Supplier ABclonal Technology Supplier No. A20891100UL
Enables identical protein binding activity Predicted to be involved in actin cytoskeleton organization endocytosis and signal transduction Located in nucleoplasm Biomarker of Huntington s disease