Learn More
Medchemexpress LLC Cipemastat | 190648-49-8 | 99.5% | 436.55 | 5 MG
SDP
Catalog No. 5000401917
Encompass_Preferred
Cipemastat is a potent, competitive inhibitor of human collagenases 1, 2, and 3, demonstrating Ki values of 3.0, 4.4, and 3.4 nM, respectively. It exhibits selectivity for these collagenases over related matrix metalloproteinases and also potently inhibits rat collagenase. In vitro studies show it inhibits IL-1α induced cartilage degradation in a concentration-dependent manner, without cytotoxic action on explant chondrocytes.
  • Potent, competitive inhibitor of human collagenases 1, 2 and 3
  • Selective for collagenases 1, 2 and 3 over related matrix metalloproteinases
  • Potently inhibits rat collagenase
  • Inhibits IL-1α induced cartilage degradation in vitro
  • Increased implanted cartilage hydroxypro-line in animal studies
  • Showed significantly greater area of cartilage in arthritic animal models
Catalog No. 50-004-01917 Supplier Medchemexpress LLC Supplier No. HY196775MG
Cipemastat is a potent, competitive inhibitor of human collagenases 1, 2, and 3, demonstrating Ki values of 3.0, 4.4, and 3.4 nM, respectively. It exhibits selectivity for these collagenases over related matrix metalloproteinases and also potently inhibits rat collagenase. In vitro studies show it inhibits IL-1α induced cartilage degradation in a concentration-dependent manner, without cytotoxic action on explant chondrocytes.
  • Potent, competitive inhibitor of human collagenases 1, 2 and 3
  • Selective for collagenases 1, 2 and 3 over related matrix metalloproteinases
  • Potently inhibits rat collagenase
  • Inhibits IL-1α induced cartilage degradation in vitro
  • Increased implanted cartilage hydroxypro-line in animal studies
  • Showed significantly greater area of cartilage in arthritic animal models