Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cipralisant | 213027-19-1 | 99.3% | 216.32 g·mol⁻¹ | C14H20N2 | 100 MG
SDP

Catalog No. 5000239870
Encompass_Preferred

Cipralisant is a potent, selective histamine H3 receptor ligand used in research to study H3 receptor pharmacology in vitro and in vivo. It is supplied as a characterized solid reagent with specified purity, formula, and storage recommendations for laboratory use.

  • Potent, selective H3 receptor ligand.
  • High purity (99.3%).
  • Molecular formula C14H20N2; molecular weight 216.32 g·mol⁻¹.
  • Soluble in DMSO ~200 mg/mL; use ultrasonic and freshly opened DMSO.
  • Store powder at -20°C (long term) or 4°C; aliquot solutions and store at -80°C to avoid freeze-thaw.

Catalog No. 50-002-39870 Supplier Medchemexpress LLC Supplier No. HY106993100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Cipralisant is a potent, selective histamine H3 receptor ligand used in research to study H3 receptor pharmacology in vitro and in vivo. It is supplied as a characterized solid reagent with specified purity, formula, and storage recommendations for laboratory use.

  • Potent, selective H3 receptor ligand.
  • High purity (99.3%).
  • Molecular formula C14H20N2; molecular weight 216.32 g·mol⁻¹.
  • Soluble in DMSO ~200 mg/mL; use ultrasonic and freshly opened DMSO.
  • Store powder at -20°C (long term) or 4°C; aliquot solutions and store at -80°C to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.