Learn More
Medchemexpress LLC Cipralisant | 213027-19-1 | 99.3% | 216.32 g/mol | C14H20N2 | 25 MG
SDP
Catalog No. 5000239933
Encompass_Preferred
Cipralisant (GT-2331) is a small-molecule histamine H3 receptor ligand used for research applications. It is reported as a potent, selective, high-affinity H3 receptor full antagonist in vivo and an agonist in vitro, with literature-reported activity values suitable for pharmacological evaluation and click-chemistry applications.
  • Potent, high-affinity histamine H3 receptor ligand (pKi ≈ 9.9).
  • Shows antagonist activity in vivo and agonist activity in vitro.
  • Reported rat H3 Ki ≈ 0.47 nM for target validation studies.
  • Contains an alkyne functional group enabling copper-catalyzed azide-alkyne click chemistry.
  • High analytical purity (reported ~99.3%), suitable for research use.
  • Supplied as a small-scale reagent for pharmacology and AD/HD research.
Catalog No. 50-002-39933 Supplier Medchemexpress LLC Supplier No. HY10699325MG
Cipralisant (GT-2331) is a small-molecule histamine H3 receptor ligand used for research applications. It is reported as a potent, selective, high-affinity H3 receptor full antagonist in vivo and an agonist in vitro, with literature-reported activity values suitable for pharmacological evaluation and click-chemistry applications.
  • Potent, high-affinity histamine H3 receptor ligand (pKi ≈ 9.9).
  • Shows antagonist activity in vivo and agonist activity in vitro.
  • Reported rat H3 Ki ≈ 0.47 nM for target validation studies.
  • Contains an alkyne functional group enabling copper-catalyzed azide-alkyne click chemistry.
  • High analytical purity (reported ~99.3%), suitable for research use.
  • Supplied as a small-scale reagent for pharmacology and AD/HD research.