Learn More
Medchemexpress LLC Cipralisant maleate | 223420-20-0 | 99.8% | 332.39 | C18H24N2O4 | 1 MG
SDP
Catalog No. 5000240005
Encompass_Preferred
Cipralisant maleate is a research-grade histamine H3 receptor ligand used in pharmacology and neuroscience research. It acts as a potent, selective H3 receptor antagonist in vivo and may display agonist activity in certain in vitro assays. The material is supplied as a high-purity solid and is intended for experimental use under appropriate laboratory conditions.
  • Potent, selective histamine H3 receptor ligand.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity (≈99.8%).
  • Stable as a powder when stored under recommended cold conditions.
  • Available in small package sizes for research, including 1 mg.
  • White to off-white solid appearance.
Catalog No. 50-002-40005 Supplier Medchemexpress LLC Supplier No. HY106993A1MG
Cipralisant maleate is a research-grade histamine H3 receptor ligand used in pharmacology and neuroscience research. It acts as a potent, selective H3 receptor antagonist in vivo and may display agonist activity in certain in vitro assays. The material is supplied as a high-purity solid and is intended for experimental use under appropriate laboratory conditions.
  • Potent, selective histamine H3 receptor ligand.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity (≈99.8%).
  • Stable as a powder when stored under recommended cold conditions.
  • Available in small package sizes for research, including 1 mg.
  • White to off-white solid appearance.