Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cipralisant maleate | 223420-20-0 | 99.8% | 332.39 g·mol-1 | C18H24N2O4 | 10 MG
SDP

Catalog No. 5000239820
Encompass_Preferred

Cipralisant maleate is a research-grade histamine H3 receptor antagonist supplied as the maleate salt. It is used in pharmacology and neuroscience studies to probe H3 receptor activity and related signaling pathways. Provided as a solid with high reported purity, this compound is for laboratory research use only and not for human or veterinary applications.

  • Orally active histamine H3 receptor antagonist for research applications.
  • High purity (99.8%).
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Molecular weight 332.39 g·mol-1; formula C18H24N2O4.
  • Stable as powder at -20°C for long-term storage; in solution store at -80°C.
  • Supplied in small laboratory-scale quantities suitable for preclinical studies.

Catalog No. 50-002-39820 Supplier Medchemexpress LLC Supplier No. HY106993A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cipralisant maleate is a research-grade histamine H3 receptor antagonist supplied as the maleate salt. It is used in pharmacology and neuroscience studies to probe H3 receptor activity and related signaling pathways. Provided as a solid with high reported purity, this compound is for laboratory research use only and not for human or veterinary applications.

  • Orally active histamine H3 receptor antagonist for research applications.
  • High purity (99.8%).
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Molecular weight 332.39 g·mol-1; formula C18H24N2O4.
  • Stable as powder at -20°C for long-term storage; in solution store at -80°C.
  • Supplied in small laboratory-scale quantities suitable for preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.