Learn More
Medchemexpress LLC Cipralisant maleate | 223420-20-0 | 99.8% | 332.39 g/mol | C18H24N2O4 | 25 MG
SDP
Catalog No. 5000239821
Encompass_Preferred
Cipralisant maleate is a potent, selective histamine H3 receptor ligand used in neuroscience research. It functions as a full antagonist in vivo and shows agonist activity in vitro, with high receptor affinity; it is provided as a solid research chemical for pharmacological and behavioral studies involving wakefulness and attention mechanisms.
  • High affinity for histamine H3 receptor.
  • Potent antagonist activity in vivo.
  • Agonist activity observed in vitro.
  • High purity suitable for research applications.
  • Available in small-scale quantities with storage recommendations for stability.
Catalog No. 50-002-39821 Supplier Medchemexpress LLC Supplier No. HY106993A25MG
Cipralisant maleate is a potent, selective histamine H3 receptor ligand used in neuroscience research. It functions as a full antagonist in vivo and shows agonist activity in vitro, with high receptor affinity; it is provided as a solid research chemical for pharmacological and behavioral studies involving wakefulness and attention mechanisms.
  • High affinity for histamine H3 receptor.
  • Potent antagonist activity in vivo.
  • Agonist activity observed in vitro.
  • High purity suitable for research applications.
  • Available in small-scale quantities with storage recommendations for stability.