Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cipralisant maleate | 223420-20-0 | 99.8% | 332.4 g/mol | C18H24N2O4 | 5 MG
SDP

Catalog No. 5000240471
Encompass_Preferred

Cipralisant maleate is the maleate salt of cipralisant, a potent and selective histamine H3 receptor ligand that functions as a full antagonist in vivo and an agonist in vitro. Supplied for research use in neuroscience and pharmacology, the compound offers high purity, good DMSO solubility, and defined storage conditions for stability.

  • Potent, selective histamine H3 receptor antagonist in vivo.
  • Agonist activity observed in vitro.
  • High reported purity (99.84%).
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Molecular weight 332.4 g/mol.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Catalog No. 50-002-40471 Supplier Medchemexpress LLC Supplier No. HY106993A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cipralisant maleate is the maleate salt of cipralisant, a potent and selective histamine H3 receptor ligand that functions as a full antagonist in vivo and an agonist in vitro. Supplied for research use in neuroscience and pharmacology, the compound offers high purity, good DMSO solubility, and defined storage conditions for stability.

  • Potent, selective histamine H3 receptor antagonist in vivo.
  • Agonist activity observed in vitro.
  • High reported purity (99.84%).
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Molecular weight 332.4 g/mol.
  • Recommended storage: powder -20°C for long term, 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.