Learn More
Medchemexpress LLC Cipralisant maleate 50mg | 223420-20-0 | 50MG
SDP
Catalog No. 5000240004
Encompass_Preferred
Cipralisant (GT-2331) (maleate) is an orally active low-toxicity potent selective high affinity histamine H3 receptor full antagonist in vivo and an agonist in vitro with a pKi of 9 9 for histamine H3 receptor and a Ki of 0 47 nM for rat histamine H3 receptor Cipralisant (maleate) has the potential for attention-deficit hyperactivity disorder research[1 [2 [3 [4 Cipralisant (maleate) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-40004 Supplier Medchemexpress LLC Supplier No. HY106993A50MG
Add to Cart
Add to Cart
Cipralisant (GT-2331) (maleate) is an orally active low-toxicity potent selective high affinity histamine H3 receptor full antagonist in vivo and an agonist in vitro with a pKi of 9 9 for histamine H3 receptor and a Ki of 0 47 nM for rat histamine H3 receptor Cipralisant (maleate) has the potential for attention-deficit hyperactivity disorder research[1 [2 [3 [4 Cipralisant (maleate) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups