Learn More
Medchemexpress LLC HY-B0664 100mg Medchemexpress, Ciprofibrate CAS:52214-84-3 Purity:>98%
SDP
Catalog No. 502082636
Encompass_Preferred
Medchemexpress, HY-B0664 100mg Ciprofibrate CAS:52214-84-3 Ciprofibrate (Win35833) is a potent peroxisome proliferator and increases the phosphorylation level of the PPARalpha. Ciprofibrate acts as an orally active hypolipidaemic agent and can be used for the research of primary hyperlipidaemias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2636 Supplier Medchemexpress LLC Supplier No. HYB0664100MG
Medchemexpress, HY-B0664 100mg Ciprofibrate CAS:52214-84-3 Ciprofibrate (Win35833) is a potent peroxisome proliferator and increases the phosphorylation level of the PPARalpha. Ciprofibrate acts as an orally active hypolipidaemic agent and can be used for the research of primary hyperlipidaemias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.