Learn More
Apexbio Technology LLC Ciprofibrate 52214-84-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062376
Encompass_Preferred
Ciprofibrate is a selective agonist targeting peroxisome proliferator-activated receptor alpha (PPAR ) a nuclear receptor involved in regulating lipid metabolism and energy homeostasis Its activation promotes gene expression linked to fatty acid oxidation triglyceride reduction and cholesterol metabolism Ciprofibrate is frequently applied as a molecular tool in biomedical research to investigate mechanisms underlying dyslipidemia atherosclerosis and metabolic-related disorders Additionally due to its receptor-specific mode of action it serves as a standard experimental reference in in vitro assays and animal models assessing lipid-lowering therapeutic strategies and PPAR -dependent signaling pathways
Catalog No. 50-000-62376 Supplier Apexbio Technology LLC Supplier No. A43065.1
Ciprofibrate is a selective agonist targeting peroxisome proliferator-activated receptor alpha (PPAR ) a nuclear receptor involved in regulating lipid metabolism and energy homeostasis Its activation promotes gene expression linked to fatty acid oxidation triglyceride reduction and cholesterol metabolism Ciprofibrate is frequently applied as a molecular tool in biomedical research to investigate mechanisms underlying dyslipidemia atherosclerosis and metabolic-related disorders Additionally due to its receptor-specific mode of action it serves as a standard experimental reference in in vitro assays and animal models assessing lipid-lowering therapeutic strategies and PPAR -dependent signaling pathways