Learn More
Medchemexpress LLC Ciprofloxacin-13C3,15N monohydrochloride | 2483830-12-0 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0356AS11MG
Encompass_Preferred
Ciprofloxacin-13C3,15N monohydrochloride is a 13C- and 15N-labeled version of Ciprofloxacin (monohydrochloride) intended for research use only. This compound serves as a valuable tool for precise tracking and quantification in various analytical methods, helping to study metabolic pathways and drug mechanisms.
  • Used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Enables precise tracking and quantification of individual atoms in metabolic pathways.
  • Minimally impacts molecular properties, aiding in distinguishing between endogenous and exogenous metabolites.
  • Reduces false positives and aids in the reconstruction of metabolic pathways.
  • Improves accuracy and reproducibility in mass spectrometry through internal standardization.
  • Facilitates non-invasive tracking of compounds and their metabolites in vivo, beneficial for pharmacometabolic kinetics studies.
Catalog No. 50-004-06270