Learn More
Medchemexpress LLC Ciprofloxacin | 85721-33-1 | MFCD00185755 | 99.8% | 1 G
SDP
Catalog No. 501874090
Encompass_Preferred
Ciprofloxacin is a potent, orally active topoisomerase IV inhibitor. It induces mitochondrial DNA and nuclear DNA damage, leading to mitochondrial dysfunction and reactive oxygen species (ROS) production. This compound exhibits anti-proliferative activity and induces apoptosis. As a fluoroquinolone antibiotic, it also possesses potent antibacterial activity.
  • Potent, orally active topoisomerase IV inhibitor
  • Induces mitochondrial and nuclear DNA damage
  • Leads to mitochondrial dysfunction and ROS production
  • Exhibits anti-proliferative activity
  • Induces apoptosis
  • Functions as a fluoroquinolone antibiotic
  • Possesses potent antibacterial activity
Catalog No. 50-187-4090 Supplier Medchemexpress LLC Supplier No. HYB03561G
Ciprofloxacin is a potent, orally active topoisomerase IV inhibitor. It induces mitochondrial DNA and nuclear DNA damage, leading to mitochondrial dysfunction and reactive oxygen species (ROS) production. This compound exhibits anti-proliferative activity and induces apoptosis. As a fluoroquinolone antibiotic, it also possesses potent antibacterial activity.
  • Potent, orally active topoisomerase IV inhibitor
  • Induces mitochondrial and nuclear DNA damage
  • Leads to mitochondrial dysfunction and ROS production
  • Exhibits anti-proliferative activity
  • Induces apoptosis
  • Functions as a fluoroquinolone antibiotic
  • Possesses potent antibacterial activity