Learn More
Enzo Life Sciences Ciprofloxacin (25g). CAS: 85721-33-1
SDP
Catalog No. 502009017
Encompass_Preferred
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria. Effective against Mycobacterium tuberculosis. Potentially anxiogenic. Alternative name: 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1- piperazinyl)-3-quinolinecarboxylic acid. Purity: ≥98% (HPLC). Solubility: Soluble in dilute hydrochloric acid; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9017 Supplier Enzo Life Sciences Supplier No. ALX380287G025
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria. Effective against Mycobacterium tuberculosis. Potentially anxiogenic. Alternative name: 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1- piperazinyl)-3-quinolinecarboxylic acid. Purity: ≥98% (HPLC). Solubility: Soluble in dilute hydrochloric acid; insoluble in water. Long Term Storage: -20°C.