Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ciprofloxacin, 98%

Catalog No. AC449620050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449620050 5 g Glass Bottle
AC449620250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449620050 Supplier Thermo Scientific Chemicals Supplier No. 449620050
  • $78.00
    $62.65 / Each of 1

    Save $15.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Ciprofloxacin, CAS# 85721-33-1, is a second-generation fluoroquinolone showing antibiotic activity commonly used against susceptible organisms mainly gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Ciprofloxacin is considered a synthetic broad-spectrum second-generation compound with antibiotic activity mainly against gram-negative bacteria
• It inhibits bacterial DNA gyrase and topoisomerase IV, enzymes that are essential for DNA replication, cell division, and causes double-strand breaks in the bacterial chromosome

Applications

• Ciprofloxacin presents more activity against gram-negative bacteria than gram-positive bacteria.
• Ciprofloxacin does not present cross-resistance between fluoroquinolones and other antibiotics classes, which is beneficial in laboratory experiments when other antibiotics are no longer effective

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85721-33-1
Molecular Formula C17H18FN3O3
Molecular Weight (g/mol) 331.34
InChI Key MYSWGUAQZAJSOK-UHFFFAOYSA-N
Synonym ciprofloxacin, ciprofloxacine, ciprobay, ciproxan, ciprofloxacina, cipro, ciprofloxacino, ciprofloxacinum, cipro iv, bacquinor
PubChem CID 2764
ChEBI CHEBI:100241
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)N4CCNCC4)F)C(=O)O

Specifications

Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Merck Index 15,2313
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in hot dmso,soluble in hot methanol
Formula Weight 331.34
Percent Purity 98%
Chemical Name or Material Ciprofloxacin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.