Learn More
MOLECULAR DEPOT LLC Ciprofloxacin BSA - 1 mg
Catalog No. 5000013518
Encompass_Preferred
Ciprofloxacin BSA conjugate is a chemical compound formed by linking ciprofloxacin an antibiotic with bovine serum albumin (BSA) This conjugate enhances solubility and stability and is used in research particularly in immunological assays For in vitro research use only
Catalog No. 50-000-13518 Supplier MOLECULAR DEPOT LLC Supplier No. B2020785
Ciprofloxacin BSA conjugate is a chemical compound formed by linking ciprofloxacin an antibiotic with bovine serum albumin (BSA) This conjugate enhances solubility and stability and is used in research particularly in immunological assays For in vitro research use only