Learn More
Medchemexpress LLC Ciprofloxacin-d8 hydrochloride | 1216659-54-9 | MFCD08705188 | 99.0% | 10 MG
SDP
Catalog No. 5000298239
Encompass_Preferred
Ciprofloxacin-d8 hydrochloride is the deuterium-labeled form of the fluoroquinolone antibiotic ciprofloxacin, supplied as the hydrochloride salt for research use. It is intended primarily as an internal standard or tracer for analytical chemistry, pharmacokinetic studies, and mass spectrometry workflows where isotopic labeling improves detection and quantitation.
  • Deuterated internal standard for LC-MS/MS assays.
  • Hydrochloride salt supplied for analytical use.
  • High isotopic and chemical purity for accurate quantitation.
  • Provided in small mg-scale quantities for method development and validation.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-98239 Supplier Medchemexpress LLC Supplier No. HYB0356S10MG
Ciprofloxacin-d8 hydrochloride is the deuterium-labeled form of the fluoroquinolone antibiotic ciprofloxacin, supplied as the hydrochloride salt for research use. It is intended primarily as an internal standard or tracer for analytical chemistry, pharmacokinetic studies, and mass spectrometry workflows where isotopic labeling improves detection and quantitation.
  • Deuterated internal standard for LC-MS/MS assays.
  • Hydrochloride salt supplied for analytical use.
  • High isotopic and chemical purity for accurate quantitation.
  • Provided in small mg-scale quantities for method development and validation.
  • For research use only; not for human or veterinary use.