Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciprofloxacin, G-Biosciences
SCI_ED

Catalog No. S100911
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S100911 5 g
S000033 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S100911 Supplier G-Biosciences Supplier No. RC970
Add to Cart
Add to Cart

Ciprofloxacin is a second generation fluoroquinolone antibiotic targeting a broad-spectrum of bacteria and mycoplasma.

It is nalidixic acid analog and has few known resistant strains. Ciprofloxacin is also effective as a plant fungicide and is sparingly soluble in water.

Chemical Identifiers

CAS 85721-33-1
Molecular Weight (g/mol) 331.34 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Gas Chromatography, GC-MS
Certifications/Compliance ISO, NIST, REACH, ISO 9001, GHS/OSHA
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 5 g
Percent Purity >98%
Grade Molecular Biology
Chemical Name or Material Ciprofloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.