Learn More
Gold Biotechnology Inc Ciprofloxacin HCl 10 g
SDP
Catalog No. NC1490707
Encompass
Ciprofloxacin is a second generation fluoroquinolone antibiotic targeting a broad-spectrum of bacteria and mycoplasma. It is nalidixic acid analog and has few known resistant strains. Ciprofloxacin is also effective as a plant fungicide. Ciprofloxacin HCl is freely soluble in water. GoldBio also offers Ciprofloxacin, a purer form of the antibiotic which does not contain HCl, but is sparingly soluble in water.
Catalog No. NC1490707 Supplier Gold Biotechnology Inc Supplier No. C86110
Ciprofloxacin is a second generation fluoroquinolone antibiotic targeting a broad-spectrum of bacteria and mycoplasma. It is nalidixic acid analog and has few known resistant strains. Ciprofloxacin is also effective as a plant fungicide. Ciprofloxacin HCl is freely soluble in water. GoldBio also offers Ciprofloxacin, a purer form of the antibiotic which does not contain HCl, but is sparingly soluble in water.