Learn More
Medchemexpress LLC HY-B0356A 500mg Medchemexpress, Ciprofloxacin (hydrochloride) CAS:93107-08-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0356A500MG
Encompass_Preferred
Medchemexpress, HY-B0356A 500mg Ciprofloxacin (hydrochloride) CAS:93107-08-5 Ciprofloxacin hydrochloride (Bay-09867 hydrochloride) is a fluoroquinolone antibiotic, exhibiting potent antibacterial activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2488