Learn More
Bioworld Ciprofloxacin Hydrochloride Solution, 93107-08-5, Sterile, 10 mg/mL
Catalog No. 50255729
Encompass_Preferred
For Mycoplasma/Bacterial Elimination from animal cell cultures.1000X, 10mg/mL, effective concentration 10ug/ml in most culture media. Mycoplasma contamination is a major problem in cell culture in biomedical and cell/molecular biology research. There is no easy visual confirmation of mycoplasma contamination which can have serious effects on key characteristics and functions of a cell line, thus compromising all results obtained using such cultures. Cipro HCl is the standard antibiotic for rapid and efficient removal of mycoplasma from contaminated cultures. Applications: Treatment of mycoplasma-infected cell lines, Decontamination of visible Pen-Strep resistant bacteria, Prevent contamination from plasmid preparation during transfections. Molecular Weight : 367.80 g/mol, Molecular Formula : C17H18FN3O3�HCl, Solubility : Soluble in water, and 0.1 M NaOH. Melting Point : 311-320 °C, 10ml
Catalog No. 50-255-729 Supplier Bioworld Supplier No. 403100311
Add to Cart
Add to Cart
For Mycoplasma/Bacterial Elimination from animal cell cultures.1000X, 10mg/mL, effective concentration 10ug/ml in most culture media. Mycoplasma contamination is a major problem in cell culture in biomedical and cell/molecular biology research. There is no easy visual confirmation of mycoplasma contamination which can have serious effects on key characteristics and functions of a cell line, thus compromising all results obtained using such cultures. Cipro HCl is the standard antibiotic for rapid and efficient removal of mycoplasma from contaminated cultures. Applications: Treatment of mycoplasma-infected cell lines, Decontamination of visible Pen-Strep resistant bacteria, Prevent contamination from plasmid preparation during transfections. Molecular Weight : 367.80 g/mol, Molecular Formula : C17H18FN3O3�HCl, Solubility : Soluble in water, and 0.1 M NaOH. Melting Point : 311-320 °C, 10ml