Learn More
Bioworld CIPROFLOXACIN HYDROCHLORIDE
Supplier:  Bioworld 403100321
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. Ciprofloxacin Hydrochloride (Powder) For Mycoplasma/Bacterial Elimination from animal cell cultures.
Catalog No. 50-255-731