Learn More
Medchemexpress LLC Ciprofloxacin hydrochloride monohydrate | 86393-32-0 | 99.7% | 5 G
SDP
Catalog No. 5000406115
Encompass_Preferred
Ciprofloxacin hydrochloride monohydrate is a potent, orally active topoisomerase IV inhibitor. It induces mitochondrial DNA and nuclear DNA damage, leading to mitochondrial dysfunction and ROS production. It also exhibits anti-proliferative activity and induces apoptosis. This compound is a fluoroquinolone antibiotic with potent antibacterial activity. This product is for research use only.
  • Potent, orally active topoisomerase IV inhibitor
  • Induces mitochondrial DNA and nuclear DNA damage
  • Leads to mitochondrial dysfunction and ROS production
  • Exhibits anti-proliferative activity
  • Induces apoptosis
  • Fluoroquinolone antibiotic with potent antibacterial activity
  • Inhibits cell proliferation and causes cell cycle arrest at G2/M phase in tendon cells
  • Shows potent activity against Y. pestis and B. anthracis
Catalog No. 50-004-06115 Supplier Medchemexpress LLC Supplier No. HYB0356B5G
Ciprofloxacin hydrochloride monohydrate is a potent, orally active topoisomerase IV inhibitor. It induces mitochondrial DNA and nuclear DNA damage, leading to mitochondrial dysfunction and ROS production. It also exhibits anti-proliferative activity and induces apoptosis. This compound is a fluoroquinolone antibiotic with potent antibacterial activity. This product is for research use only.
  • Potent, orally active topoisomerase IV inhibitor
  • Induces mitochondrial DNA and nuclear DNA damage
  • Leads to mitochondrial dysfunction and ROS production
  • Exhibits anti-proliferative activity
  • Induces apoptosis
  • Fluoroquinolone antibiotic with potent antibacterial activity
  • Inhibits cell proliferation and causes cell cycle arrest at G2/M phase in tendon cells
  • Shows potent activity against Y. pestis and B. anthracis