Learn More
Cayman Chemical CIprostencalcIum salt 10mg
SDP
Catalog No. 502967701
Encompass_Preferred
Ciprostene is the 9β-methyl analog of carbaprostacyclin and a stable analog of PGI2. Ciprostene exhibits biological activity similar to PGI2, but is 30-fold less potent. In patas monkeys, ciprostene induces hypotension and causes tachycardia when administered at a dose of 0.16 UG/kg/min.{4373} In addition, ciprostene inhibits ADP-induced platelet aggregation ex vivo and in vitro with ID50 values of 9.1 UG/kg/min and 60 ng/ml, respectively.{4373,4375}
Catalog No. 50-296-7701 Supplier Cayman Chemical Supplier No. 1821610 MG
Ciprostene is the 9β-methyl analog of carbaprostacyclin and a stable analog of PGI2. Ciprostene exhibits biological activity similar to PGI2, but is 30-fold less potent. In patas monkeys, ciprostene induces hypotension and causes tachycardia when administered at a dose of 0.16 UG/kg/min.{4373} In addition, ciprostene inhibits ADP-induced platelet aggregation ex vivo and in vitro with ID50 values of 9.1 UG/kg/min and 60 ng/ml, respectively.{4373,4375}