Learn More
TARGETMOL CHEMICALS INC Ciproxifan maleate 10MG
SDP
Catalog No. 502879450
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ciproxifan maleate (FUB 359)(FUB-359 maleate) is a highly potent and selective histamin H3-receptor antagonist with IC50 of 9.2 nM, with low apparent affinity at other receptor subtypes. Purity 99.45%
Catalog No. 50-287-9450 Supplier TARGETMOL CHEMICALS INC Supplier No. T200410MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ciproxifan maleate (FUB 359)(FUB-359 maleate) is a highly potent and selective histamin H3-receptor antagonist with IC50 of 9.2 nM, with low apparent affinity at other receptor subtypes. Purity 99.45%