Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ciproxifan maleate | 184025-19-2 | C20H22N2O6 | 5 MG
SDP

Catalog No. 5000372034
Encompass_Preferred

Ciproxifan maleate is a potent, selective, orally bioavailable, and competitive antagonist of histamine H3-receptor, with an IC50 of 9.2 nM. It displays low apparent affinity at other receptor subtypes and can be used for the research of aging disorders and Alzheimer's disease.

  • Purity: 99.68%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 386.40
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: 3.57 mg/mL
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-72034 Supplier Medchemexpress LLC Supplier No. HY152895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ciproxifan maleate is a potent, selective, orally bioavailable, and competitive antagonist of histamine H3-receptor, with an IC50 of 9.2 nM. It displays low apparent affinity at other receptor subtypes and can be used for the research of aging disorders and Alzheimer's disease.

  • Purity: 99.68%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 386.40
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: 3.57 mg/mL
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.