Learn More
Medchemexpress LLC Ciproxifan maleate | 184025-19-2 | C20H22N2O6 | 50 MG
SDP
Catalog No. 5000373177
Encompass_Preferred
Ciproxifan maleate is a potent, selective, orally bioavailable, and competitive antagonist of the histamine H3-receptor, with an IC50 of 9.2 nM. It shows low apparent affinity at other receptor subtypes and can be used for research of aging disorders and Alzheimer's disease.
  • Potent and selective antagonist of histamine H3-receptor
  • Orally bioavailable
  • Useful for research on aging disorders and Alzheimer's disease
  • Inhibits [3H]HA release from rat cerebral cortex synaptosomes
  • Improves accuracy in five-choice tasks in rats
  • Induces neocortical electroencephalogram activation in cats
  • Exhibits oral bioavailability in mice
Catalog No. 50-003-73177 Supplier Medchemexpress LLC Supplier No. HY1528950MG
Ciproxifan maleate is a potent, selective, orally bioavailable, and competitive antagonist of the histamine H3-receptor, with an IC50 of 9.2 nM. It shows low apparent affinity at other receptor subtypes and can be used for research of aging disorders and Alzheimer's disease.
  • Potent and selective antagonist of histamine H3-receptor
  • Orally bioavailable
  • Useful for research on aging disorders and Alzheimer's disease
  • Inhibits [3H]HA release from rat cerebral cortex synaptosomes
  • Improves accuracy in five-choice tasks in rats
  • Induces neocortical electroencephalogram activation in cats
  • Exhibits oral bioavailability in mice