Learn More
Medchemexpress LLC Ciproxifan (maleate) | 184025-19-2 | MFCD06411563 | 99.7% | 386.40 g/mol | C20H22N2O6 | 10 MG
SDP
Catalog No. 5000287052
Encompass_Preferred
Ciproxifan maleate is a potent, selective, orally bioavailable competitive antagonist of the histamine H3 receptor supplied as a white to off-white powder for research use. It is used as an analytical standard and research reagent in neuroscience and aging-disease studies, with reported nanomolar potency.
  • Potent H3 receptor antagonist (IC50 ≈ 9.2 nM)
  • High purity (≈ 99.7%)
  • Provided as a powdered maleate salt for stability and handling
  • Suitable for neuroscience and Alzheimer's disease research
  • Available in small pack sizes for analytical and screening use
  • Includes CAS number 184025-19-2 and molecular weight 386.40 g/mol
Catalog No. 50-002-87052 Supplier Medchemexpress LLC Supplier No. HY1528910MG
Ciproxifan maleate is a potent, selective, orally bioavailable competitive antagonist of the histamine H3 receptor supplied as a white to off-white powder for research use. It is used as an analytical standard and research reagent in neuroscience and aging-disease studies, with reported nanomolar potency.
  • Potent H3 receptor antagonist (IC50 ≈ 9.2 nM)
  • High purity (≈ 99.7%)
  • Provided as a powdered maleate salt for stability and handling
  • Suitable for neuroscience and Alzheimer's disease research
  • Available in small pack sizes for analytical and screening use
  • Includes CAS number 184025-19-2 and molecular weight 386.40 g/mol