Learn More
Selleck Chemical LLC Ciproxifan Maleate S2813-5mg
SDP
Catalog No. 503112460
Encompass_Preferred
Ciproxifan (FUB 359) is a highly potent and selective histamin H3-receptor antagonist with IC50 of 9 2 nM with low apparent affinity at other receptor subtypes
Catalog No. 50-311-2460 Supplier Selleck Chemical LLC Supplier No. S28135MG
Ciproxifan (FUB 359) is a highly potent and selective histamin H3-receptor antagonist with IC50 of 9 2 nM with low apparent affinity at other receptor subtypes