Learn More
Cayman Chemical CIproxIfnmaleate 10mg
SDP
Catalog No. 502988011
Encompass_Preferred
A histamine H3 receptor antagonist (Ki = 0.5 nM for inhibition of histamine release in rat synaptosomal preparations); selective for histamine H3 over histamine H1 and H2, M3 muscarinic, and α1D- and β1-adrenergic receptors (Kis = 3.15-25 µM), as well as 5-HT1B, 5-HT2A, 5-HT3, and 5-HT4 (Kis = 1.58-15 µM) in radioligand binding assays; inhibits R-(–)-α-methylhistamine-induced relaxation in precontracted isolated guinea pig ileal longitudinal muscle from 3-300 nM; reduces R-(–)-α-methylhistamine-induced water consumption in rats (ED50 = 0.09 mg/kg); increases wake episode duration and latency to fall asleep in rats when administered at a dose of 2 mg/kg
Catalog No. 50-298-8011 Supplier Cayman Chemical Supplier No. 2951310 MG
A histamine H3 receptor antagonist (Ki = 0.5 nM for inhibition of histamine release in rat synaptosomal preparations); selective for histamine H3 over histamine H1 and H2, M3 muscarinic, and α1D- and β1-adrenergic receptors (Kis = 3.15-25 µM), as well as 5-HT1B, 5-HT2A, 5-HT3, and 5-HT4 (Kis = 1.58-15 µM) in radioligand binding assays; inhibits R-(–)-α-methylhistamine-induced relaxation in precontracted isolated guinea pig ileal longitudinal muscle from 3-300 nM; reduces R-(–)-α-methylhistamine-induced water consumption in rats (ED50 = 0.09 mg/kg); increases wake episode duration and latency to fall asleep in rats when administered at a dose of 2 mg/kg