Learn More
Grainger HONEYWELL, Circular Paper Chart, Honeywell
Catalog No. NC2384975
Encompass_Preferred
The Honeywell 5MEC7 is a circular paper chart recorder for use with Honeywell temperature, humidity, pressure, and flow controllers. It features a 7 day rotation period with 24 hour circumferential grid lines and dual pen capability to monitor two variables at once. The chart drive accepts multiple input signals with ranges from 1 to 5 VDC, 4 to 20 mA, or 10 to 50 mA. It has an accuracy of ±0.5% of full scale. The 5MEC7 comes with mounting hardware, pens, ink and a roll of chart paper. It can be panel, surface or pipe mounted. This circular chart recorder provides a continuous hardcopy record of process measurements.
Catalog No. NC2384975 Supplier Grainger Supplier No. 5MEP9
The Honeywell 5MEC7 is a circular paper chart recorder for use with Honeywell temperature, humidity, pressure, and flow controllers. It features a 7 day rotation period with 24 hour circumferential grid lines and dual pen capability to monitor two variables at once. The chart drive accepts multiple input signals with ranges from 1 to 5 VDC, 4 to 20 mA, or 10 to 50 mA. It has an accuracy of ±0.5% of full scale. The 5MEC7 comes with mounting hardware, pens, ink and a roll of chart paper. It can be panel, surface or pipe mounted. This circular chart recorder provides a continuous hardcopy record of process measurements.