Learn More
Medchemexpress LLC Ciraparantag (PER977) | 1438492-26-2 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1866010MG
Encompass_Preferred
Ciraparantag (PER977) is a small synthetic cationic molecule used as a research reagent to reverse a variety of anticoagulants. It antagonizes direct factor Xa and thrombin inhibitors, as well as low-molecular-weight and unfractionated heparins; it does not reverse vitamin K antagonists or argatroban. Supplied in powder and solution formats for laboratory use (research use only).
  • Broad-spectrum reversal of direct oral anticoagulants and heparins.
  • Active against direct factor Xa and thrombin inhibitors.
  • Does not reverse vitamin K antagonists or argatroban.
  • Available in powder and solution formats.
  • Intended for in vitro and preclinical research applications.
Catalog No. 50-002-94731