Learn More
Medchemexpress LLC HY-101300 5mg Medchemexpress, Cirazoline (hydrochloride) CAS:40600-13-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1013005MG
Encompass_Preferred
Medchemexpress, HY-101300 5mg Cirazoline (hydrochloride) CAS:40600-13-3 Cirazoline hydrochloride (LD 3098 hydrochloride) is a potent competitive full α1A-adrenergic receptor (α1A-AR) agonist (Ki=120 nM) and only a partial agonist at α1B-AR (Ki= 960 nM) and α1D-AR (Ki=660 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0626