Learn More
Aviva Systems Biology Corp CIRBP Antibody - N-terminal region (ARP40349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CIRBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CIRBP. Alternate names: CIRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-57404