Learn More
Proteintech Group Inc CIRBP Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 102092AP150UL
Encompass_Preferred
The CIRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CIRBP. This antibody recognizes human, mouse, rat antigen. The CIRBP antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Catalog No. 50-225-6358