Learn More
Fine Science Tools Circle Forceps, 0.7 mm, Stainless Steel, 11.5 cm
SDP
Catalog No. NC1588529
Encompass
Cupped forceps for atraumatic gripping of organs and tissues.Tip Diameter: 0.7; Alloy / Material: Stainless Steel; Length: 11.5cm
Catalog No. NC1588529 Supplier Fine Science Tools Supplier No. 1107211
Cupped forceps for atraumatic gripping of organs and tissues.Tip Diameter: 0.7; Alloy / Material: Stainless Steel; Length: 11.5cm