Learn More
LGC GENOMICS LLC (Lucigen) CircLigase ssDNA Ligase, 1,000 U @ 100 U/L
Catalog No. NC9439450
Encompass
CircLigase™ ssDNA Ligase is a thermostable ATP-dependent ligase that catalyzes intramolecular ligation (i.e. circularization) of ssDNA templates having a 5-phosphate and a 3-hydroxyl group
This item is not returnable. View return policy
Catalog No. NC9439450 Supplier LGC GENOMICS LLC Supplier No. CL4111K
CircLigase™ ssDNA Ligase is a thermostable ATP-dependent ligase that catalyzes intramolecular ligation (i.e. circularization) of ssDNA templates having a 5-phosphate and a 3-hydroxyl group