Learn More
LGC GENOMICS LLC (Lucigen) CircLigase II ssDNA Ligase, 1,000 U @ 100 U/LAn ATP-independent, non-catalytic thermostable ligase that catalyses the intramolecular ligation (i.e. circularisation) of ssDNA and ssRNA templates.
Catalog No. NC9869616
Encompass
CircLigase™ II ssDNA Ligase is a thermostable enzyme that catalyzes intramolecular ligation (i.e. circularization) of ssDNA templates having a 5-phosphate and a 3-hydroxyl group
This item is not returnable. View return policy
Catalog No. NC9869616 Supplier LGC GENOMICS LLC Supplier No. CL9021K
CircLigase™ II ssDNA Ligase is a thermostable enzyme that catalyzes intramolecular ligation (i.e. circularization) of ssDNA templates having a 5-phosphate and a 3-hydroxyl group