Learn More
Grainger Circuit Breaker 250A 3P 600VAC JG
Catalog No. 502750850
Encompass_Preferred
The Grainger Approved 250A 3P Circuit Breaker is a 3-pole circuit breaker with a 250 ampere interrupting rating. It has a voltage rating of 600VAC and is designed for use in panelboards, switchboards, and motor control centers. This thermal-magnetic circuit breaker provides overload and short circuit protection. It has a width of 3.75 inches and features line and load screw type terminals. The circuit breaker is UL listed and CSA certified.
Catalog No. 50-275-0850 Supplier Grainger Supplier No. 14G983
Add to Cart
Add to Cart
Request a Quote
The Grainger Approved 250A 3P Circuit Breaker is a 3-pole circuit breaker with a 250 ampere interrupting rating. It has a voltage rating of 600VAC and is designed for use in panelboards, switchboards, and motor control centers. This thermal-magnetic circuit breaker provides overload and short circuit protection. It has a width of 3.75 inches and features line and load screw type terminals. The circuit breaker is UL listed and CSA certified.