Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Circuit Breaker 30A Plug In 120V 1P

Supplier:  Grainger 15F782

Encompass_Preferred

Circuit Breaker 30A Plug In 120V 1P

Catalog No. 50-275-3955


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.