Learn More
Grainger Circuit Breaker 60A 3P 600VAC HG
Catalog No. 502750909
Encompass_Preferred
The Grainger Approved 1-Pole 15 Amp 120/277 Volt DIN Rail Mount Circuit Breaker is a thermal-magnetic circuit breaker designed for overload and short-circuit protection. It has a maximum voltage rating of 277VAC and a continuous current rating of 15A. This circuit breaker features a 1-pole design and mounts onto 35mm DIN rails. It has a 10kA interrupting rating. The circuit breaker's connections are screw-type terminals that accept wire sizes from #14-#10 AWG. It conforms to IEC 60947-2 standards. This DIN rail mount breaker provides reliable circuit protection for industrial control panels and machinery.
Catalog No. 50-275-0909 Supplier Grainger Supplier No. 14G975
The Grainger Approved 1-Pole 15 Amp 120/277 Volt DIN Rail Mount Circuit Breaker is a thermal-magnetic circuit breaker designed for overload and short-circuit protection. It has a maximum voltage rating of 277VAC and a continuous current rating of 15A. This circuit breaker features a 1-pole design and mounts onto 35mm DIN rails. It has a 10kA interrupting rating. The circuit breaker's connections are screw-type terminals that accept wire sizes from #14-#10 AWG. It conforms to IEC 60947-2 standards. This DIN rail mount breaker provides reliable circuit protection for industrial control panels and machinery.