Learn More
Cole-Parmer Extech CB10 AC Circuit Breaker Finder/Receptacle Tester
Supplier:  Cole-Parmer 2002117
Encompass
The AC circuit breaker finder/receptable tester allows you to quickly and safely located 100 to 125 volt AC circuit breakers and fuses with audible and visual indicators. Features include a variable sensitivity adjustment to pinpoint correct circuit breakers, push button GFCI test, and convenient storage position for transmitter/receptacle tester.
Catalog No. 50-992-963