Learn More
BRAIN RESEARCH LABORATORIES CIRCULAR GLASS DISH 125 MM
Catalog No. NC2383526
Encompass
Pyrex® Crystallizing Dishes.Pyrex® 7740 Borosilicate Glass.125mm diameter X 65mm high, 740mlFor laboratory use only.
Catalog No. NC2383526 Supplier BRAIN RESEARCH LABORATORIES Supplier No. 1256
Pyrex® Crystallizing Dishes.Pyrex® 7740 Borosilicate Glass.125mm diameter X 65mm high, 740mlFor laboratory use only.