Learn More
Cole-Parmer O C White EM-45-5D-W Circular Illuminated Magnifier, 5" Dia, 2.25x Mag, 5 Diopters, White; Clamp Base
Catalog No. NC1422349 Shop All Cole-Parmer Products
Encompass
These metal shade magnifiers feature a 5" diameter glass lens. The 22-watt fluorescent bulb emits a high quality light for your inspection needs. Maintain desired position of 45" (1.1 m) arm reach with internal spring design. Shade handle offers a safe and convenient method of adjusting magnifier position. Operates on 120 VAC.
Catalog No. NC1422349 Supplier Cole-Parmer Supplier No. 4180772
These metal shade magnifiers feature a 5" diameter glass lens. The 22-watt fluorescent bulb emits a high quality light for your inspection needs. Maintain desired position of 45" (1.1 m) arm reach with internal spring design. Shade handle offers a safe and convenient method of adjusting magnifier position. Operates on 120 VAC.