Learn More
Perkin Elmer US LLC Demountable Cell Window - KBr, 4 mm, Qty. 2
Catalog No. 509043344
Encompass_Preferred
Circular Demountable Windows are available in a variety of window materials. Samples are placed on one window and the second window is pressed lightly on top of the first. Spacers of various thicknesses can be used to vary the cell path length. These windows require, but do not include, the Universal Demountable Cell (L1270986).
Catalog No. 50-904-3344 Supplier Perkin Elmer US LLC Supplier No. L1271202
Circular Demountable Windows are available in a variety of window materials. Samples are placed on one window and the second window is pressed lightly on top of the first. Spacers of various thicknesses can be used to vary the cell path length. These windows require, but do not include, the Universal Demountable Cell (L1270986).