Learn More
Grainger DICKSON, Circular Paper Chart, Dickson
Catalog No. NC2229215
Encompass_Preferred
The Dickson Circular Paper Chart Recorder is a 7-day circular chart recorder that continuously monitors and records temperature or humidity. It has a 6 inch diameter chart that rotates once per week and can record temperatures from -40 to 160°F or humidity from 10 to 95% RH. The recorder comes with a capillary pen arm and uses standard 12 hour or 24 hour charts. It features a plastic case and clear acrylic lens and can be wall mounted. The recorder requires minimal maintenance and provides a simple way to get permanent temperature and humidity records.
This item is not returnable. View return policy
Catalog No. NC2229215 Supplier Grainger Supplier No. 1LXK9
The Dickson Circular Paper Chart Recorder is a 7-day circular chart recorder that continuously monitors and records temperature or humidity. It has a 6 inch diameter chart that rotates once per week and can record temperatures from -40 to 160°F or humidity from 10 to 95% RH. The recorder comes with a capillary pen arm and uses standard 12 hour or 24 hour charts. It features a plastic case and clear acrylic lens and can be wall mounted. The recorder requires minimal maintenance and provides a simple way to get permanent temperature and humidity records.