Learn More
Grainger Circular Saw Blade 10 in Blade 80 Teeth
Catalog No. 502750243
Encompass_Preferred
The DEWALT 10-Inch Circular Saw Blade is a 10 inch diameter blade with 24 teeth. It has a 5/8 inch arbor size and is 0.095 inches thick. The blade is made from high grade carbide tipped steel for long life and smooth cuts. It features DEWALT's Precision Computer Balanced Plate Technology which reduces vibration and noise during use. This general purpose blade can cut through wood, plywood, chipboard and more. It provides fast, clean cuts in a variety of materials.
Catalog No. 50-275-0243 Supplier Grainger Supplier No. 14K257
The DEWALT 10-Inch Circular Saw Blade is a 10 inch diameter blade with 24 teeth. It has a 5/8 inch arbor size and is 0.095 inches thick. The blade is made from high grade carbide tipped steel for long life and smooth cuts. It features DEWALT's Precision Computer Balanced Plate Technology which reduces vibration and noise during use. This general purpose blade can cut through wood, plywood, chipboard and more. It provides fast, clean cuts in a variety of materials.